Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
arpc1b
Entrez Gene ID
335474
Gene Name
actin related protein 2/3 complex, subunit 1B
Gene Aliases
arpc1a, cb871, fj15h02, wu:fj08e12, wu:fj15h02, zgc:64056
Chromosome Location
Chr.3: 40809048 - 40820479 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213156.1 NP_998321.1 4 - 5 508 74
GenBank mRNA BC053229.1 - 4 - 5 508 74
BC164463.1 - 3 - 4 426 74

View Full Product Details