Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
slit1a
Entrez Gene ID
556123
Gene Name
slit homolog 1a (Drosophila)
Gene Aliases
-
Chromosome Location
Chr.13: 555089 - 638485 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034968.1 NP_001030140.1 3 - 4 468 82
GenBank mRNA BC163538.1 - 3 - 4 360 82
DQ177286.1 - 3 - 4 468 82

View Full Product Details