Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tnfrsf21
Entrez Gene ID
564874
Gene Name
tumor necrosis factor receptor superfamily, member 21
Gene Aliases
dr6, im:6795346, wu:fa55e01, wu:fb02e11, wu:fc29a09, wu:fi27h08
Chromosome Location
Chr.20: 35799912 - 35841581 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042688.2 NP_001036153.2 4 - 5 1795 68
GenBank mRNA BC141795.1 - 4 - 5 1795 68
DQ812118.1 - 4 - 5 1466 68

View Full Product Details