Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
scn12aa
Entrez Gene ID
795688
Gene Name
sodium channel, voltage gated, type XII, alpha a
Gene Aliases
-
Chromosome Location
Chr.2: 51887312 - 52045385 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044922.1 NP_001038387.1 25 - 26 4572 141
GenBank mRNA DQ149507.1 - 25 - 26 4572 141

View Full Product Details