Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
syt4
Entrez Gene ID
335357
Gene Name
synaptotagmin IV
Gene Aliases
wu:fj05d11, zgc:73240
Chromosome Location
Chr.7: 23907746 - 23916118 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199948.1 NP_956242.1 2 - 3 1076 84
GenBank mRNA BC059580.1 - 2 - 3 1076 84

View Full Product Details