Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
laptm4a
Entrez Gene ID
100003844
Gene Name
lysosomal protein transmembrane 4 alpha
Gene Aliases
wu:fb57c07, wu:fj99e08
Chromosome Location
Chr.20: 43775793 - 43786433 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214685.2 NP_999850.2 2 - 3 295 94
GenBank mRNA BC058878.1 - 2 - 3 322 94
BC155748.1 - 2 - 3 295 94

View Full Product Details