Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clcn1a
Entrez Gene ID
795133
Gene Name
chloride channel, voltage-sensitive 1a
Gene Aliases
si:dkey-14o18.5
Chromosome Location
Chr.19: 9611877 - 9663055 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082910.1 NP_001076379.1 1 - 2 151 92

View Full Product Details