Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cpsf4
Entrez Gene ID
30203
Gene Name
cleavage and polyadenylation specific factor 4
Gene Aliases
nar
Chromosome Location
Chr.3: 40289387 - 40295257 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131009.1 NP_571084.1 3 - 4 598 85
XM_021469299.1 XP_021324974.1 3 - 4 633 85
GenBank mRNA BC045289.1 - 3 - 4 601 85
BC164456.1 - 3 - 4 339 85
U70479.1 - 3 - 4 598 85

View Full Product Details