Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
chp1
Entrez Gene ID
325361
Gene Name
calcineurin-like EF-hand protein 1
Gene Aliases
chp, si:dkey-41n20.1, wu:fc69b04, zgc:63904
Chromosome Location
Chr.13: 45035790 - 45063613 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199715.1 NP_956009.1 5 - 6 569 104
GenBank mRNA BC054566.1 - 5 - 6 569 104

View Full Product Details