Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
vps26b
Entrez Gene ID
406684
Gene Name
VPS26, retromer complex component B
Gene Aliases
wu:fb49d07, wu:fb50d06, zgc:56297
Chromosome Location
Chr.15: 227600 - 234026 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213375.1 NP_998540.1 2 - 3 655 129
XM_005157511.4 XP_005157568.1 3 - 4 588 129
GenBank mRNA BC046056.1 - 2 - 3 655 129

View Full Product Details