Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ssrp1a
- Entrez Gene ID
-
386844
- Gene Name
- structure specific recognition protein 1a
- Gene Aliases
-
ssrp1, wu:fa11b03, wu:fd10f06
- Chromosome Location
-
Chr.14: 174031 - 200429 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_212802.2 |
NP_997967.2 |
11 - 12 |
1457 |
90 |
GenBank mRNA |
BC056311.1 |
- |
11 - 12 |
1363 |
90 |
- Gene Symbol
- ssrp1b
- Entrez Gene ID
-
449949
- Gene Name
- structure specific recognition protein 1b
- Gene Aliases
-
im:7140964
- Chromosome Location
-
Chr.1: 44710943 - 44739559 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
CT642102.2 |
- |
2 - 3 |
208 |
90 |
View Full Product Details