Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
elavl2
Entrez Gene ID
431719
Gene Name
ELAV like neuron-specific RNA binding protein 2
Gene Aliases
zgc:91918
Chromosome Location
Chr.22: 38252726 - 38274235 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002172.2 NP_001002172.2 7 - 8 965 60
GenBank mRNA BC072716.1 - 7 - 8 921 60

View Full Product Details