Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mcfd2
Entrez Gene ID
449676
Gene Name
multiple coagulation factor deficiency 2, ER cargo receptor complex subunit
Gene Aliases
hm:zeh1186, wu:fc23b10, wu:fd50d08, zgc:103713, zgc:109915
Chromosome Location
Chr.13: 8685081 - 8692854 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005939.1 NP_001005939.1 3 - 4 452 106
GenBank mRNA BC083471.1 - 3 - 4 452 106
BC163979.1 - 2 - 3 372 106

View Full Product Details