Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
krcp
Entrez Gene ID
323406
Gene Name
kelch repeat-containing protein
Gene Aliases
wu:fb97e08, wu:fi38d02, zgc:123032
Chromosome Location
Chr.7: 24881660 - 24888634 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012366.1 NP_001012366.1 9 - 10 1291 78
XM_005172254.4 XP_005172311.1 9 - 10 1367 78
GenBank mRNA AY835383.1 - 9 - 10 1291 78
BC108043.1 - 9 - 10 1355 78

View Full Product Details