Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
atp6v0a1b
Entrez Gene ID
553691
Gene Name
ATPase H+ transporting V0 subunit a1b
Gene Aliases
wu:fj37e01, zgc:112214
Chromosome Location
Chr.24: 7594105 - 7632140 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020666.1 NP_001018502.1 3 - 4 308 116
XM_009297243.2 XP_009295518.1 3 - 4 345 116
GenBank mRNA BC095691.1 - 3 - 4 308 116

View Full Product Details