Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sema3aa
Entrez Gene ID
30266
Gene Name
sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3Aa
Gene Aliases
sema1a, sema3a1, semaZ1a
Chromosome Location
Chr.4: 7414 - 61242 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131060.1 NP_571135.1 16 - 17 2225 72
GenBank mRNA AF086761.1 - 16 - 17 2225 72
BC162381.1 - 16 - 17 1938 72
BC162398.1 - 16 - 17 1938 72
CU458855.1 - 16 - 17 1868 72

View Full Product Details