Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
stt3a
Entrez Gene ID
266797
Gene Name
STT3 oligosaccharyltransferase complex catalytic subunit A
Gene Aliases
cb437, itm1, wu:fb38c11
Chromosome Location
Chr.10: 39212898 - 39233144 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201458.2 NP_958866.1 4 - 5 382 66
GenBank mRNA BC046072.1 - 4 - 5 383 66
BC063234.1 - 4 - 5 343 66

View Full Product Details