Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hm13
Entrez Gene ID
259190
Gene Name
histocompatibility (minor) 13
Gene Aliases
H13, SPP, cb228, wu:fc11a06, zgc:56660, zgc:86862
Chromosome Location
Chr.23: 480433 - 493392 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212572.2 NP_997737.2 6 - 7 808 72
XM_005161890.4 XP_005161947.1 6 - 7 845 72
GenBank mRNA BC053414.1 - 6 - 7 72
BC071503.1 - 6 - 7 808 72

View Full Product Details