The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014345.2 | NP_001014367.2 | 13 - 14 | 1644 | 78 |
GenBank mRNA | AY880251.1 | - | 13 - 14 | 1644 | 78 |
BC091981.1 | - | 9 - 10 | 985 | 78 | |
BC164709.1 | - | 8 - 9 | 981 | 78 |