Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
golim4b
Entrez Gene ID
541535
Gene Name
golgi integral membrane protein 4b
Gene Aliases
im:7146775, im:7156643, zgc:113641
Chromosome Location
Chr.2: 36701241 - 36721297 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014349.2 NP_001014371.2 5 - 6 764 101
XM_005171289.4 XP_005171346.1 5 - 6 845 101
GenBank mRNA BC091983.1 - 5 - 6 768 101

View Full Product Details