Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
skib
Entrez Gene ID
30113
Gene Name
v-ski avian sarcoma viral oncogene homolog b
Gene Aliases
fc17g02, wu:fc17g02, wu:fk79g04
Chromosome Location
Chr.11: 396572 - 404947 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130938.1 NP_571013.1 2 - 3 1345 79
GenBank mRNA AF060118.1 - 2 - 3 1345 79

View Full Product Details