Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
oxt
Entrez Gene ID
352920
Gene Name
oxytocin
Gene Aliases
IT-NP, fj33a01, itnp, oxtl, wu:fj33a01, wu:fq40d10
Chromosome Location
Chr.5: 72087619 - 72093255 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178291.2 NP_840076.1 3 - 4 390 71
GenBank mRNA AY069956.1 - 3 - 4 390 71

View Full Product Details