Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scfd1
Entrez Gene ID
317732
Gene Name
sec1 family domain containing 1
Gene Aliases
SI:dZ115O7.1, SI:dZ234G15.3, SI:dZ234G15.4, SI:dZ234G15.5, SI:dZ234G15.7, dZ234G15.2, si:busm1-234g15.2, si:dz234g15.2, sly1
Chromosome Location
Chr.17: 28771027 - 28797395 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182861.1 NP_878281.1 4 - 5 333 127
GenBank mRNA AY167023.1 - 4 - 5 333 127
BC050950.1 - 4 - 5 309 127
BC163864.1 - 4 - 5 306 127
BC163870.1 - 4 - 5 306 127

View Full Product Details