Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fam49bb
Entrez Gene ID
404610
Gene Name
family with sequence similarity 49 member Bb
Gene Aliases
si:dkeyp-105g12.2, zgc:76981
Chromosome Location
Chr.24: 10793398 - 10897320 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207078.1 NP_996961.1 11 - 12 1075 97
GenBank mRNA BC066542.1 - 11 - 12 1075 97
BC152197.1 - 11 - 12 1139 97

View Full Product Details