Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
aldh3b1
Entrez Gene ID
282559
Gene Name
aldehyde dehydrogenase 3 family, member B1
Gene Aliases
aldh3d1, cb508, fb17d04, wu:fb17d04
Chromosome Location
Chr.3: 34120184 - 34128952 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173221.2 NP_775328.2 5 - 6 598 118
XM_005163981.4 XP_005164038.1 5 - 6 642 118
GenBank mRNA AF254955.1 - 4 - 5 492 118
BC049338.1 - 5 - 6 598 118

View Full Product Details