Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rpl13
Entrez Gene ID
378961
Gene Name
ribosomal protein L13
Gene Aliases
zgc:92272
Chromosome Location
Chr.7: 67467615 - 67476495 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198143.1 NP_937786.1 3 - 4 424 116
XM_005166666.4 XP_005166723.1 4 - 5 569 116
GenBank mRNA AF385081.1 - 3 - 4 424 116
AY561516.1 - 4 - 5 468 116
BC075977.1 - 4 - 5 455 116

View Full Product Details