Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lin7a
Entrez Gene ID
393682
Gene Name
lin-7 homolog A (C. elegans)
Gene Aliases
lin7b, zgc:73090
Chromosome Location
Chr.4: 21751762 - 21851488 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200709.1 NP_957003.1 2 - 3 249 95
XM_005164866.4 XP_005164923.1 2 - 3 268 95
GenBank mRNA AY584747.1 - 2 - 3 248 95
BC059460.1 - 2 - 3 249 95

View Full Product Details