Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pcolce2b
Entrez Gene ID
393432
Gene Name
procollagen C-endopeptidase enhancer 2b
Gene Aliases
zgc:63590
Chromosome Location
Chr.24: 5871589 - 5889763 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200460.1 NP_956754.1 5 - 6 772 124
GenBank mRNA BC055153.1 - 5 - 6 772 124

View Full Product Details