Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
c8g
Entrez Gene ID
393837
Gene Name
complement component 8, gamma polypeptide
Gene Aliases
zgc:77874
Chromosome Location
Chr.24: 17031129 - 17039617 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200863.1 NP_957157.1 4 - 5 566 127
GenBank mRNA BC062865.1 - 4 - 5 566 127

View Full Product Details