Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
sept7a
Entrez Gene ID
394136
Gene Name
septin 7a
Gene Aliases
cdc10, wu:fb38h06, zgc:56383
Chromosome Location
Chr.19: 11331815 - 11425610 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201161.1 NP_957455.1 6 - 7 549 120
XM_021468025.1 XP_021323700.1 7 - 8 675 120
XM_021468026.1 XP_021323701.1 6 - 7 680 120
XM_021468027.1 XP_021323702.1 7 - 8 675 120
XM_021468028.1 XP_021323703.1 6 - 7 680 120
GenBank mRNA BC049418.1 - 6 - 7 549 120
KC513820.1 - 6 - 7 120

View Full Product Details