Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
kin
Entrez Gene ID
555316
Gene Name
Kin17 DNA and RNA binding protein
Gene Aliases
fc49f10, si:ch211-235a22.3, wu:fc49f10, zgc:123169
Chromosome Location
Chr.4: 25181572 - 25188103 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030086.2 NP_001025257.2 1 - 2 315 136
GenBank mRNA BC109449.1 - 1 - 2 315 136

View Full Product Details