Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
retsatl
Entrez Gene ID
677660
Gene Name
retinol saturase (all-trans-retinol 13,14-reductase) like
Gene Aliases
RetSat B, wu:fi35b01, zgc:123334
Chromosome Location
Chr.9: 44994253 - 44998722 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040004.2 NP_001035093.2 3 - 4 712 114

View Full Product Details