Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ghrhrl
Entrez Gene ID
793334
Gene Name
growth hormone releasing hormone receptor, like
Gene Aliases
ghrhr
Chromosome Location
Chr.23: 40073451 - 40126974 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082482.1 NP_001075951.1 7 - 8 827 100
GenBank mRNA DQ991247.1 - 7 - 8 827 100

View Full Product Details