Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
prkcbp1l
Entrez Gene ID
323479
Gene Name
protein kinase C binding protein 1, like
Gene Aliases
im:7138336, wu:fb99d12, zgc:63775
Chromosome Location
Chr.23: 9809630 - 9863632 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199641.1 NP_955935.1 8 - 9 887 84
XM_005161943.4 XP_005162000.1 9 - 10 996 84
XM_005161944.4 XP_005162001.1 9 - 10 1458 84
XM_005161945.4 XP_005162002.1 9 - 10 1301 84
XM_005161946.4 XP_005162003.1 9 - 10 917 84
XM_009296662.3 XP_009294937.1 9 - 10 896 84
XM_009296664.3 XP_009294939.1 9 - 10 899 84
XM_009296665.3 XP_009294940.1 9 - 10 996 84
GenBank mRNA BC058064.1 - 8 - 9 887 84
BC091798.1 - 9 - 10 1384 84

View Full Product Details