Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lysmd4
Entrez Gene ID
100149102
Gene Name
LysM, putative peptidoglycan-binding, domain containing 4
Gene Aliases
si:cr925803.2, si:dkey-111m11.2, zgc:77861
Chromosome Location
Chr.18: 234181 - 244650 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200850.2 NP_957144.2 2 - 3 366 122
GenBank mRNA BC062528.1 - 2 - 3 320 122
CF347270.1 - 2 - 3 318 122
CK682157.1 - 2 - 3 363 122
CK869133.1 - 2 - 3 377 122
CO795793.1 - 2 - 3 370 122
CO930564.1 - 2 - 3 379 122

View Full Product Details