The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002403.1 | NP_001002403.1 | 4 - 5 | 380 | 146 |
GenBank mRNA | BC076179.1 | - | 4 - 5 | 380 | 146 |