Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
jmjd7
Entrez Gene ID
550278
Gene Name
jumonji domain containing 7
Gene Aliases
zgc:110265
Chromosome Location
Chr.17: 10680032 - 10738049 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017615.1 NP_001017615.1 3 - 4 618 72
XM_021467157.1 XP_021322832.1 3 - 4 666 72
GenBank mRNA BC092834.1 - 3 - 4 618 72
BC164846.1 - 3 - 4 493 72

View Full Product Details