Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ankhb
Entrez Gene ID
323738
Gene Name
ANKH inorganic pyrophosphate transport regulator b
Gene Aliases
ankh, wu:fc08d03
Chromosome Location
Chr.2: 31957615 - 32006312 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_194370.1 NP_919351.1 8 - 9 1283 94
GenBank mRNA AJ302035.1 - 8 - 9 1283 94
BC081575.1 - 8 - 9 94

View Full Product Details