Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
jade3
Entrez Gene ID
322209
Gene Name
jade family PHD finger 3
Gene Aliases
phf16, wu:fb53c02
Chromosome Location
Chr.6: 37478765 - 37513010 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201652.2 NP_970614.2 11 - 12 1760 84
GenBank mRNA BC045468.1 - 11 - 12 1760 84
BC165248.1 - 9 - 10 1581 84

View Full Product Details