Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
zfpm2a
Entrez Gene ID
568996
Gene Name
zinc finger protein, FOG family member 2a
Gene Aliases
FOG-A, fog2a, zgc:152696
Chromosome Location
Chr.16: 43818892 - 44127431 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039635.1 NP_001034724.1 6 - 7 922 141
XM_009292598.3 XP_009290873.1 6 - 7 968 141
GenBank mRNA AY515851.1 - 6 - 7 866 141
BC162670.1 - 6 - 7 845 141
DQ015976.1 - 6 - 7 922 141

View Full Product Details