Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tnnt3a
Entrez Gene ID
58084
Gene Name
troponin T type 3a (skeletal, fast)
Gene Aliases
tnnt
Chromosome Location
Chr.25: 31405184 - 31418457 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131565.1 NP_571640.1 10 - 11 692 129
XM_005163151.3 XP_005163208.3 11 - 12 832 129
GenBank mRNA AF180889.1 - 10 - 11 692 129
BC053304.1 - 11 - 12 715 129

View Full Product Details