The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_174862.3 | NP_777287.2 | 5 - 6 | 1249 | 78 |
XM_005156971.4 | XP_005157028.1 | 6 - 7 | 1582 | 78 | |
XM_005156972.4 | XP_005157029.1 | 7 - 8 | 1756 | 78 | |
XM_005156973.4 | XP_005157030.1 | 6 - 7 | 1269 | 78 | |
GenBank mRNA | AF342950.1 | - | 3 - 4 | 539 | 78 |
AF349413.3 | - | 6 - 7 | 1268 | 78 | |
AJ275911.3 | - | 6 - 7 | 1313 | 78 | |
AJ414566.1 | - | 6 - 7 | 1269 | 78 | |
BC086848.1 | - | 5 - 6 | 1249 | 78 | |
BC164792.1 | - | 5 - 6 | 1224 | 78 |