Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cbwd
Entrez Gene ID
406536
Gene Name
COBW domain containing
Gene Aliases
fb76b04, fc73a04, wu:fb76b04, wu:fc73a04, zgc:77617
Chromosome Location
Chr.8: 29378 - 53722 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213253.1 NP_998418.1 6 - 7 567 89
GenBank mRNA BC065429.1 - 6 - 7 567 89

View Full Product Details