Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dact1
Entrez Gene ID
405799
Gene Name
dishevelled-binding antagonist of beta-catenin 1
Gene Aliases
dapper1, dpr1, frd1, frodo1
Chromosome Location
Chr.17: 11005873 - 11015878 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214731.3 NP_999896.3 3 - 4 904 146
GenBank mRNA AY521628.1 - 3 - 4 605 146
AY549443.1 - 3 - 4 587 146
BC127382.1 - 3 - 4 904 146
BC165174.1 - 3 - 4 639 146

View Full Product Details