Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nhej1
Entrez Gene ID
406593
Gene Name
nonhomologous end-joining factor 1
Gene Aliases
zgc:85657
Chromosome Location
Chr.6: 7085895 - 7108613 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214749.1 NP_999914.1 2 - 3 422 99
GenBank mRNA BC068348.1 - 2 - 3 422 99

View Full Product Details