Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
her6
Entrez Gene ID
30288
Gene Name
hairy-related 6
Gene Aliases
HER-6, cb129
Chromosome Location
Chr.6: 193858 - 195543 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131079.2 NP_571154.2 1 - 1 146 152
GenBank mRNA BC059551.1 - 1 - 1 114 152
EH450285.1 - 1 - 1 155 152
EH579523.1 - 1 - 1 162 152
X97333.1 - - 144 152

View Full Product Details