Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dub
Entrez Gene ID
566846
Gene Name
duboraya
Gene Aliases
zgc:153303
Chromosome Location
Chr.21: 22917981 - 22951659 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045422.1 NP_001038887.1 6 - 6 854 143
XM_005161193.4 XP_005161250.1 5 - 5 812 143
GenBank mRNA BC122224.1 - 6 - 6 854 143
BC164256.1 - 5 - 5 801 143

View Full Product Details