Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
aqp10b
Entrez Gene ID
100034395
Gene Name
aquaporin 10b
Gene Aliases
si:dkey-204a24.5
Chromosome Location
Chr.19: 7809723 - 7816886 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083047.1 NP_001076516.1 6 - 6 942 146
XM_005159392.4 XP_005159449.1 6 - 6 1296 146

View Full Product Details