Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nkd3l
Entrez Gene ID
100034538
Gene Name
NKD inhibitor of WNT signaling pathway 3, like
Gene Aliases
si:dkey-98l21.2
Chromosome Location
Chr.19: 33368152 - 33370271 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083083.1 NP_001076552.1 2 - 2 747 95

View Full Product Details