Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lrmda
Entrez Gene ID
553730
Gene Name
leucine rich melanocyte differentiation associated
Gene Aliases
c10orf11, c13h10orf11, si:ch211-39f22.1, zgc:112463
Chromosome Location
Chr.13: 16920792 - 17468023 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020701.2 NP_001018537.1 7 - 7 1346 78
XM_021480580.1 XP_021336255.1 8 - 8 4930 78
GenBank mRNA BC095822.1 - 7 - 7 1349 78

View Full Product Details